Learn More
UC Biodevices Corporation KANAMYCIN DETECTION ELISA TEST
Supplier: UC Biodevices Corporation 5501E212
KANAMYCIN DETECTION ELISA TEST - Kit Contents1) Microtiter plate ( 8 wells×12) precoated with antibodies to mouse IgG.2) Non-coated Dilution Microwells (8 wells×12 strips)3) 5×kanamycin standard solutions (1ml/each): 0 ng/ml, 0.2 ng/ml, 0.5ng/ml,1.0ng/ml, 3.0ng/ml. (Note: 1mg of kanamycin=775U of kanamycin, so that1U/ml=1.29μg/ml=1290ppb)4) KAN Enzyme Conjugate(1×) (15ml)5) Washing buffer(10×) (50ml)6) KAN dilution buffer (2x50ml)7) Substrate TMB solution (17ml) 8) Stop solution (7ml)9) Instruction Manual
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.