Learn More
Strategic Applications Inc Micro500 K3 EDTA 500uL Blood Tube - Plasma, Push Cap, 100/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Strategic Applications Inc M500E3
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
500ul K3 EDTA Micro500 Blood Tube, Purple Cap. SAI's Micro500 blood sampling tubes replicate the dimensions of the discontinued BD Microtainer for 500ul blood collections. With the same slim 8 mm X 43 mm size, no adjustments for methods or instrumentation are required. 100/pack. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply. Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.