Learn More
Biorex Labs K-Check Controls for serum, plasma/blood ketones determination
Supplier:  Biorex Labs B1777
Encompass_Preferred
Recommended CONTROLS for Serum/Plasma or Blood ketones determination when using K-CHECK REAGENT TABLETS. Each set of 3 vials include a 5 ml each of Negative, Low positive and a High positive control. Each box has 2 sets (total of 6 vials, 5 ml each). 30 days expiration after opening a vial. 1 year expiration for an unopened vial. Made with simulated serum matrix, free of potential infectious agents and free of animal or human derived products.
Catalog No. NC1551709
Explore available promotions