Learn More
Biorex Labs Reagent Tablet K-Check Ketone 100 per Bottle
Supplier: Biorex Labs B1555
K-CHECK tablets are intended to determine the qualitative/semi-quantitative presence of two Ketone bodies i.e. acetone and acetoacetic acid in Urine, Serum, Plasma or Blood. 100 tablets/bottle.