Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ JW 67
Wnt pathway inhibitor; induces degradation of active β-catenin
$242.44 - $1010.36
Specifications
Purity | >97% |
---|---|
Product Type | JW 67 |
Molecular Weight (g/mol) | 394.38 |
Inhibitors | JW 67 |
Description
Inhibitor of canonical Wnt pathway signaling (IC50 = 1.17 μM); targets the β-catenin destruction complex (GSK-3β/AXIN/APC) to induce β-catenin degradation. Selective for the canonical Wnt pathway over the Sonic hedgehog (Shh) and NF-κB pathways. Blocks G1/S cell cycle progression in colorectal cancer (CRC) cell lines (GI50 = 7.8 μM).Specifications
>97% | |
JW 67 | |
394.38 | |
JW 67 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title