Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ VaporTrap™ Filter
SCI_ED

Reduces harmful VOC vapors found inside safety cabinets.

Supplier:  Justrite™ 29916/EMD

Catalog No. S04776MF


Add to cart

Description

Description

  • Stainless-steel wire mesh cartridge containing activated carbon absorbs volatile organic chemical vapors
  • Cylindrical design allows 360° air flow for maximum surface contact
  • High-strength magnets offer flexible placement and repositioning anywhere within cabinet
  • Date notation area on label serves as a reminder for replacement
  • Variable life cycle based upon vapor types and concentrations
  • Magnetized 4 × 5.5 in. (10.2 × 13.9cm) base for easy attachment

Specifications

Specifications

1.58 kg
Vapor trap filter
8.75 in.
VOC vapors inside safety cabinet
22.2 cm
4 x 5.5 in. Magnetized base, Stainless Steel wire mesh cartridge containing activated Carbon medium
3.5 lb.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit