Learn More
Justrite™ Cease-Fire™ Waste Receptacles
The safe answer to waste paper, cigarette ash and butts, and other solvent-free combustibles accumulation
$212.43 - $610.79
Specifications
Certifications/Compliance FM, UL
Color Gray
Quantity 1 Ea.
Material Steel, Aluminum
Product Type Waste Receptacle
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Height (English) Weight (English) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Height (English) Weight (English) Price Quantity & Availability  
19-822-304
View Documents
Justrite™
26655G
55 gal. 24 in. 35.5 in. 39 lb.
Each for $610.79
Add to Cart
19-088215
View Documents
Justrite™
26626G
6 gal. 12.12 in. 17.75 in. 3 lb.
Each for $212.43
Add to Cart
Description
Description
  • Specially contoured to direct smoke and gas from any fire inside the container, back into the combustion area
  • Cut off the air supply and extinguishing flames in seconds
  • Provide around-the-clock protection against trash fires
  • Should be filled three-fourth full only
  • Steel body with aluminum head
  • Black in color
Cigarette Ash and Butt Receptacle
  • Grill guard keeps out other kinds of trash for safe containment and extinguishing of lit cigarettes
Suitable for use in plants, hospitals, schools, offices and stores
Certifications
FM; UL for cigarette receptacle
Specifications
Specifications
FM, UL
1 Ea.
Waste Receptacle
Gray
Steel, Aluminum