Learn More
Cytiva Jumbosep™ Centrifugal Device Membrane Inserts - 3K membrane insert, gray (12/pkg)
Supplier: Cytiva OD003C65
Convenient and Reliable Concentration, Purification, and Diafiltration of 20 to 60 mL of Biological Samples-Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes-Achieve 50X concentration and > 90% recovery in just minutes-Low protein-binding Omega™ membrane and polysulfone housing minimize losses due to non-specific binding-Versatile Omega membrane is available in a variety of MWCOs-Color-coded and laser etched for easy identification -Built-in deadstop prevents spinning to dryness-Unique sealing mechanism prevents retentate leakage and filtrate contamination-Economical - Sample reservoir and filtrate receiver can be sanitized or autoclaved, and reused- Omega (modified polyethersulfone) membrane Insert sold separately This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.