Learn More
Kurin Jet (Blood Culture Collection Device)
SDP
Jet is a blood culture (BC) collection device which significantly lowers BC contamination rates. Jet automatically sidelines the initial 0.15 mL of blood, which may contain contaminants from the patient’s own skin, ensuring that the best possible blood specimen reaches the collection bottles.
Supplier:  Kurin TJPIV12
 View more versions of this product
Catalog No. 23-232-023
Due to product restrictions, please to purchase or view availability for this product.
Explore available promotions
Description
Description
  • Lowers blood culture contamination rate
  • Helps achieve <1% contamination rate
  • Minimal blood volume waste
  • Closed System - All-In-One Device
  • Fits BD, bioMerieux & ThermoFisher Blood Culture Bottles
Specifications
Specifications
Jet for IV Connection for ThermoFisher Blood Culture Bottles (case)
Blood Culture Collection
200 per Cs.
IV Connection, Kurin Jet, Vacutainer Shield to fit Thermo BC Bottles