Learn More
Med Vet International Sterile IV Administration Set, 20dr/mL, 72" with Injection Site
Supplier: Med Vet International IV20
Our IV administration sets feature universal kink-resistant tubing with smooth action roller clamp, and Y injection site. 20 drops per mL rate. Compatible with both bottles and bags. Roller clamp controls flow rate of intravenous medication or fluids. Sterile Meets a wide range of solution administration requirements. Tubing Length: 72"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.