Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Isovaleric Acid
SCI_ED

Click to view available options
Quantity:
25 mL
Packaging:
Solution
This item is not returnable. View return policy
503-74-2
C5H10O2
102.13
MFCD00002726
GWYFCOCPABKNJV-UHFFFAOYSA-N
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
10430
CHEBI:28484
3-methylbutanoic acid
CC(C)CC(O)=O
This item is not returnable. View return policy
503-74-2
C5H10O2
102.13
MFCD00002726
GWYFCOCPABKNJV-UHFFFAOYSA-N
isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
10430
CHEBI:28484
3-methylbutanoic acid
CC(C)CC(O)=O

This product is an Educational Grade Chemical — no C of A is available.

Density 0.9370g/mL
Assay Percent Range 99 to 100% w/v
Packaging Solution
Linear Formula (CH3)2CHCH2COOH
Quantity 25 mL
Formula Weight 102.07
Chemical Name or Material Isovaleric Acid

ShelfLife : 2 years

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit