Learn More
Fisherbrand™ Isotemp™ Stirrer
Experience precise stirring control and exceptional safety performance for your routine protocols. Available in three sizes, which provide flexibility from microscale chemistries to production operations.
Description
Experience precise stirring control and exceptional safety performance for your routine protocols. Available in three sizes, which provide flexibility from microscale chemistries to production operations.
Advanced technology
- Slow-speed stirring, consistent speed control, and strong magnetic coupling
Flexible, robust design
- Top plate choices in ceramic or aluminum
- Raised display design protects electronics from spills
- Optional splash guard protection shield
- Variable sizes for different sample volumes
- Includes stir bar
Certifications
cCSAus (100-120V models), CE (230V models)
Compliance
RoHS compliant
Warranty and Services
Three years
Specifications
50 to 1500 rpm | |
230 V | |
4.25 x 4.25 in. | |
230V 50/60Hz | |
UK/EU/CN |
Ceramic | |
10 x 5 x 3.6 in. (25.4 x 12.7 x 9.1cm) | |
10.8 x 10.8 cm | |
50/60 Hz | |
Stirrer |