Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators, Solid-Stainless Steel Door PROMO
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
Specifications
23 cu. ft. | |
2°C to 10°C | |
R290 | |
1 | |
4 Full | |
45 kg | |
cULus | |
Microprocessor | |
2 years parts and labor, plus 3 year compressor warranty | |
Key Lock | |
32 x 27.75 x 81 in.(813 x 705 x 2057 mm) | |
NEMA 5-15 P | |
392 lb. |
651 L | |
5°C | |
Solid | |
Stainless Steel | |
100 lb. | |
Painted White Steel | |
White | |
1 Rear access port | |
RPSA | |
28.5 x 24 x 61.75 in. (724 x 610 x 1569 mm) | |
5.8 A | |
115 V 60 Hz | |
178 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction