Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators are designed with your needs in mind.
Description
- Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
- Internal plug/outlet
- Special safety sensor turns the internal plug/outlet off if a hydrocarbon (coolant) leak is detected
Specifications
49 cu. ft. | |
2°C to 10°C | |
R290 | |
2 | |
100 lb./full shelf; 50 lb./half shelf | |
Painted White Steel | |
White | |
1 Rear access port and 2 side access ports | |
CPGA | |
28.5 x 49 x 61.75 in. (724 x 1245 x 1569 mm) | |
8.8 A | |
115 V 60 Hz | |
271 kg |
1387 L | |
5°C | |
Glass | |
8 Full, 2 Half | |
45 kg/full shelf, 22.5 kg/half shelf | |
cULus, ENERGYSTAR | |
Microprocessor | |
2 years parts and labor, plus 3 year compressor warranty | |
Key Lock | |
32 x 52.7 x 81 in.(813 x 1339 x 813 mm) | |
NEMA 5-15 P | |
597 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction