Learn More
Advanced Chemical Sensors Isopropyl Alcohol Vapor Monitor
Supplier: Advanced Chemical Sensors OV57
ACS Vapor Monitor can be worn as a badge near the breathing zone to measure personal exposure, or placed in a room to measure area concentration.