Learn More
Mystaire™ Isola™ Pro Filtered Workstation
SDP
Chemical-resistant construction with superior filtration. Isola™ Pro filtered workstations are fitted with the EverSafe™ III TOUCH advanced microprocessor safety controller.
Supplier:  Mystaire™ MYISL48P
Request Bulk Order
 View more versions of this product
Catalog No. 15-338-904
Add to Cart
Description
Description
  • EverSafe™ III TOUCH touch screen controller constantly monitors fume hood performance with audible and visible alarms
  • Vividly displayed status of face velocity in linear feet per minute or meters per second
  • 18-Icon display for simple touch control of hood features and functions
  • LED lighting for true illumination of the entire work area
  • Three-stage filtration to confirm to SEFA 9-2010 DHIII recommendations
  • Single-wall, thermally-fused structural polypropylene construction for superior chemical resistance
  • Variable airflow control with automatic airflow sensor calibration
  • Solid-state gas detection with three sensitivity set points
  • Conforms to UL, CSA and CE electrical requirements
  • Integral polypropylene spill basin with rolled foil entry
  • 110 or 220 VAC
  • Built-in filter and application validation
  • Optional installation of polypropylene sink
Specifications
Specifications
Filtered Workstation
Polypropylene
28 in.
627.38 x 1206.5 x 934.72 mm (24.7 x 47.5 x 36.8 in.)
9 in.
Bonded Carbon and/or HEPA Options
60 to 100 lfm
711.2 mm
53 in.
1346.2 mm
Product Suggestions
Product Suggestions