Description
- EverSafe™ III TOUCH touch screen controller constantly monitors fume hood performance with audible and visible alarms
- Vividly displayed status of face velocity in linear feet per minute or meters per second
- 18-Icon display for simple touch control of hood features and functions
- LED lighting for true illumination of the entire work area
- Three-stage filtration to confirm to SEFA 9-2010 DHIII recommendations
- Single-wall, thermally-fused structural polypropylene construction for superior chemical resistance
- Variable airflow control with automatic airflow sensor calibration
- Solid-state gas detection with three sensitivity set points
- Conforms to UL, CSA and CE electrical requirements
- Integral polypropylene spill basin with rolled foil entry
- 110 or 220 VAC
- Built-in filter and application validation
- Optional installation of polypropylene sink
Specifications
Filtered Workstation | |
Polypropylene | |
28 in. | |
627.38 x 901.7 x 934.72 mm (24.7 x 35.5 x 36.8 in.) | |
9 in. |
Bonded Carbon and/or HEPA Options | |
60 to 100 lfm | |
711.2 mm | |
53 in. | |
1346.2 mm |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov