Learn More
Mystaire™ Isola™ Filtered Workstation Gas Phase Filters
SDP
For use with Isola™ filtered workstations. Provides operator safety from potentially harmful fumes and odors.
Supplier:  Mystaire™ MY4114
 View more versions of this product
Catalog No. 15-338-945
Add to Cart
Description
Description
  • Manufactured with high-grade coconut shell carbon
  • Activated carbon filter that provides more surface area for a higher degree of absorbency
  • Chemically impregnated carbon for increased load capacity
Specifications
Specifications
Mineral Acids
3.5 in.
20 in.
88.9 mm
508 mm
Gas Phase Filter
Isola Filtered Workstation (48 in.)
34 lb.
18 in.
508 x 457.2 x 88.9 mm (20 x 18 x 3.5 in.)
457.2 mm
Product Suggestions
Product Suggestions