Learn More
Mystaire™ Isola™ Filtered Workstation Gas Phase Filters
For use with Isola™ filtered workstations. Provides operator safety from potentially harmful fumes and odors.
Supplier: Mystaire™ MY4104
Description
- Manufactured with high-grade coconut shell carbon
- Activated carbon filter that provides more surface area for a higher degree of absorbency
- Chemically impregnated carbon for increased load capacity
Specifications
Hydrocarbons | |
3.5 in. | |
24 in. | |
88.9 mm | |
609.6 mm | |
Gas Phase Filter |
Isola Filtered Workstation (48 in.) | |
34 lb. | |
18 in. | |
508 x 457.2 x 88.9 mm (20 x 18 x 3.5 in.) | |
457.2 mm | |
Carbon |