Learn More
LI-COR IRDye 800CW NHS Ester (0.5 mg)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: LI-COR 92970020
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
IRDye infrared dyes from LI-COR Biosciences are available for researchers developing applications using near-infrared (NIR) fluorescence. These unique dyes are used in a variety of NIR imaging applications including Western blotting, plate-based assays, protein arrays, tissue section imaging, and molecular activity measurements.Standard NHS ester chemistry is used to produce custom probes labeled with LI-COR IRDye infrared dyes. The NHS ester reactive group provides the functionality for labeling primary and secondary amines, such as lysine residues in proteins.Properties: Chemical Formula: C50H54N3Na3O17S4; Molecular Weight: 1166.20 g/mol; Exact Mass: 1165.20
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.