Learn More
Cole-Parmer Digi-Sense Professional Dual-Laser Infrared Thermometer with Bluetooth Connectivity, 50:1
Supplier: Cole-Parmer 2025025
Using the free Digi-Sense D/S IR Therm app, this dual-laser infrared (IR) thermometer with Bluetooth technology allows you to transmit data directly to your Android or iOS device. View data at a safe distance from dangerous equipment or hazardous locations and use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file. Use the app to attach photos and notes to the records creating a clear reference point for your workideal for record keeping at large sites. The IR thermometer offers fast response time and high accuracy due to its 50 to 1 distance-to-sight ratio. Dual targeting lasers provide precise indication of the measurement area. It is ideal for a wide range of applications including food preparation, safety and fire inspection, plastic molding, marine, HVAC, and fleet maintenance.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.