Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Light Blue Flame Resistant Henley
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
3X-Large (Long)
4X-Large
Large
Large (Long)
Medium
Small
X-Large
X-Large (Long)
Description
- This CAT 2 AR/FR, NFPA™ 2112 compliant, lightweight henley is a casual, comfortable and stylish part of your FR system.
- With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better.
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Button
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m&superscript_2;)
- Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
- Closure: Four-button placket
- Pocket: One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Features: Moisture-wicking, breathable fabric, Raglan sleeves with full underarm gussets providing extra mobility, Hidden rib knit cuffs to keep sleeves in place
- Product Collection: iQ Series Comfort, iQ Series
- Packaging: Polybag
Specifications
Size | 4X-Large |
Gender | Men's |
Material | Cotton, Polyester, POD |
Color | Light Blue |
Neck Style | Henley |
Sleeve Style | Raglan Sleeve |
Construction | Knit |
Closure Type | Button |
Disposable | Reusable |
No. of Pockets | 2 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction