Learn More
Regis Technologies Inc Ion Pairing Reagents and Buffers, Regis, ION PAIR CONCENTRATE Q8 (100 mL) Pack of 1
SDP
Supplier:  Regis Technologies Inc 1404038200
Encompass_Preferred
Q-Series Ion Pair Concentrates (For Anions) are Regis Quaternary Amines, Bulk Ion Pair Reagents (for Anions). The Q-Series is comprised of quaternary alkyltriethylamines that can be used for the resolution of negatively charged species. This unique set of cationic reagents was developed to complement the Sulfonate Series (S-Series) and is exclusively manufactured by Regis. The Quaternary Alkyltriethylamines are available as premixed 0.5 M solutions of alkyl amines. When diluted to 1 L with HPLC-grade water, a 10 mL bottle forms a 0.005 M buffered solution.
Catalog No. 50-442-962