Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodoplatinate spray reagent | 100mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences I9157100ML
Iodoplatinate spray reagent is suitable for the detection of alkaloids,[1] sulfur[2] amines and organic nitrogen containing compounds on Thin Layer Chromatography (TLC) plates.