Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine, 0.0282 N Solution, For Residual Chlorine, APHA, Spectrum™ Chemical
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
Specifications
CAS | 7553-56-2, 7732-18-5 |
Quantity | 4 L |
Molecular Formula | I2 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
SMILES | II |
IUPAC Name | diiodine |
Molecular Weight (g/mol) | 253.81 |
Concentration or Composition (by Analyte or Components) | 0.0282 N ± 0.0003 N |
Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction