Learn More
Invitrogen™ E-Gel™ EX Agarose Gels, 1%
Precast, fast-resolving gels with 11 wells and an openable format
$207.00 - $343.00
Specifications
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers |
---|---|
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Requires: Require the E-Gel iBase™ Power System for running and the E-Gel Safe Imager™ Real-Time Transilluminator for real-time viewing of sample migration.
Description
- Contain a proprietary fluorescent nucleic acid stain (excitation: 490nm, emission: 520nm) that can be viewed with blue light, which minimizes DNA damage and allows detection down to 1ng/band of DNA
- Complete separation (by molecular weight) in 10 minutes
- Live monitoring of DNA migration without the hazards of UV light
- Easy access to the gel for extraction of DNA bands or transfer to membranes for Southern blotting
- Demonstrate over five-fold greater sensitivity than comparable gels containing ethidium bromide allows less sample use per run
- Fast analysis of RNA samples to check on their integrity before proceeding with downstream application
- The gel cassette is designed to open with a Gel Knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis
- Single comb well format
- Sample Type: Double-Stranded DNA (dsDNA), Single-Stranded RNA
- Sample Volume; 20μL
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers | |
E-Gel | |
20 μL | |
SYBR™ Gold | |
Molecular Weight | |
Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA | |
Single Comb |
1% | |
EX Agarose Gel | |
100 to 5000 bp | |
11-well | |
Preparative without dyes, Non-preparative | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.