Learn More
Investigating Animal Behavior
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Study the response of organisms to environmental conditions.
Supplier: Neo/Sci Corporation 202933
Includes: Eight choice chambers and prepared solutions to test various environmental conditions; additional choice chambers available
Description
In this lab investigation, students use specially molded choice chambers to study the response of organisms to environmental conditions.
- Determine the environmental preference of locally available animals, such as sow bugs, ants, earthworms and other small invertebrates
- Test students' predictions regarding habitat preference, as well as the innate and learned behaviors of animals
Specifications
Investigate Animal Behavior Lab | |
Students use specially molded choice chambers to study the response of organisms to environmental conditions | |
Eight choice chambers and prepared solutions to test various environmental conditions, additional choice chambers available | |
40 Students |