Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Motic™ Swift Line Inverted Infinity and Phase Contrast Microscope

Motic Swift Line Inverted Infinity and Phase Contrast Microscope boasts infinity corrected and phase optics, LED illumination, trinocular head, and ELWD (Extra Long Working Distance) 0.30 N.A. condenser for larger samples.
Supplier: Motic™ Swift Line AE31R
Description
- Trinocular head Siedentopf type with light splitter (100% or 20/80%)
- Ergonomic 45° observation angle
- Side-facing quintuple nosepiece
- Infinity corrected optics: 4X infinity, 10X and 20X infinity and phase
- ELWD N.A. 0.30 condenser
- LED illumination quartz halogen 6 V/30 W with intensity control
- Plain stage with glass and metal inserts
Specifications
40X, 100X, 200X | |
Research, University, Laboratory, Advanced Science | |
AE-31R |
Infinity and Phase | |
LED illumination |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title