Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIARDIA II™
Used to detect Giardia lamblia in stool specimens. State-of-the-art enteric disease antibody-based ELISA test whose method is standardized and reproducible to minimize variability, and which correlates highly with IFA-confirmed microscopy. Developed and Manufactured by TECHLAB, Inc.
Supplier: Techlab Inc 30405
This item is not returnable.
View return policy
Description
- Test yields results in less than two hours
- Assay can be run on the benchtop or automated with Dynex automated systems
- Break-apart microplate wells minimize reagent use
- Works with any stool consistency
Specifications
Giardia Cyst Antigen | |
96-well Plate | |
Conjugate, diluent, positive control, wash solution, stop solution, substrate, microassay plate | |
96 Tests |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction