Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purus™ Intro Mat Replacement Sticky Mat: PROMO

Decrease the amount of dust and dirt that typically tracks in from foot-traffic and equipment wheels.

Supplier:  Purus™ PS 2430 34 GPM

Catalog No. 19-125-802PM

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.

View alternate product


Description

Description

  • The Intro Mat system is comprised of a 30 layered mat applied to a reusable polystyrene frame (not included)
  • Each layer of the mat is coated with an adhesive that is formulated to capture soil and contain it until the layer is peeled away and discarded
  • Nonskid backing applied to the bottom of the frame provides stability on carpets, tile, concrete, linoleum, wood and other floor surfaces
  • The frame mat can be easily relocated to high traffic areas and removed during routine floor maintenance procedures
  • Gray
  • L x W: 30 x 24 in. 
  • 30 layers per pad
  • 4 sticky mats per case
Specifications

Specifications

Protect workplace, office and home floors from dirt abrasion and contamination
30 x 24 in.
Film: 35μm; Adhesive: 8μm
Film: 1.4 mil, Adhesive: 0.3 mil
LDPE Film
Gray
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit