Learn More
Chemglass Life Sciences Internal Bushing, #15, PTFE, 1/4" NPT, #110 Viton O-Ring
Supplier: Chemglass Life Sciences CG36442
Internal Bushing, #15, PTFE, 1/4" NPT, #110 Viton O-Ring. New front seal PTFE bushing with female NPT for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings.