Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Interactive Whiteboard CDs: Earth Science, High School
Illustrated review questions for use on Interactive Whiteboard or computer.
Specifications
Compatibility | Windows™, Mac™ |
---|---|
Format | CD-ROM |
For Use With (Application) | Perfect for use in review and assessment preparation by an individual student or for use on an Interactive Whiteboard with the entire class |
Includes | 750 Interactive review cards |
License | Site |
Catalog Number | Mfr. No. | Category | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Category | Description | Price | Quantity & Availability | ||||
S93997B
|
NewPath Learning
549010 |
Chemistry | Chemistry Review Interactive Whiteboard | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
S93994B
|
NewPath Learning
549007 |
Biology | Biology Review Interactive Whiteboard | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
S93995B
|
NewPath Learning
549008 |
Earth Science | Earth Science Review Interactive Whiteboard | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Each CD features 750 illustrated, interactive review cards for use on an Interactive Whiteboard or computer. Display questions from a single or multiple topics and save as lesson plans. Provides instant feedback along with time on task, number of attempts, percent answered correctly and number of questions remaining.
Some of the topics covered include:
- The Science of Earth Science
- Maps as Models of the Earth
- The History of Earth
- Earth's Crust
Specifications
Windows™, Mac™ | |
Perfect for use in review and assessment preparation by an individual student or for use on an Interactive Whiteboard with the entire class | |
Site |
CD-ROM | |
750 Interactive review cards | |
9 to 12 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title