Learn More
Hach Pacific Pty Ltd Intellical MTC101 Low Maintenance Gel Filled ORP/RedOx Electrode
Intellical MTC101 is a digital combination oxidation reduction potential (ORP/RedOx) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor.
$621.50
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
50-199-8053
|
Hach Pacific Pty Ltd
MTC10101 |
Standard/Lab |
Each of 1 for $621.50
|
|
|||||
Description
Intellical MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor. MTC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. Provides fast, stable response in a variety of applicationsUltimate traceability in measurement historyUser alerted when re-calibration is neededCan be moved between meters without the need to re-calibrate or re-enter measurement settings
Intellical MTC101 is a digital combination oxidation reduction potential (ORP/RedOx) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor. MTC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. Laboratory and field models available. Intellical MTC101 provides ultimate traceability in measurement history, user alert when re-calibration is needed, ability to be moved between meters without the need to re-calibrate or re-enter measurement settings.
Standard/laboratory version:
- Suitable for use in wastewater, drinking water and aqueous media applications
- Cable length 1 or 3m
- Shockproof
- Not suitable for use with organic solvents
Rugged/field version:
- Suitable for use in external environments such as rivers, surface and ground waters, ponds, lakes, sea, drinking water reservoir for water quality, environmental and treatment process purposes
- Cable length 5, 10, 15 or 30m; Stainless steel body
- Yellow coloured cable onto which you can clip depth markers
- Waterproof up to 30 meters for 24 hours (IP68 rating)
Specifications
Single/Open |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.