Learn More
Abbott Integrated E-Z Split Key Cup
Integrated E-Z split key cup rapid screening test for simultaneous, qualitative detection of analytes in human urine
$271.00 - $323.00
Specifications
Certifications/Compliance | CLIA waived |
---|---|
Sample Type | Human Urine |
Test Time | 5 min. |
Catalog Number | Mfr. No. | Detectable Analytes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Price | Quantity & Availability | |||||
22-911-143
|
Abbott
DOA3257019 |
Cocaine (COC), MET, OPI, Phencyclidine (PCP), THC (Cannabinoids) |
Pack of 25 for $271.00
|
|
|||||
22-911-144
|
Abbott
DOA1167019 |
AMP, MET, OPI, Phencyclidine (PCP), THC (Cannabinoids), Cocaine (COC) |
Pack of 25 for $323.00
|
|
|||||
Description
- Detects drugs specified at or above cutoffs
- Built in test control
- Built in Specimen Validity Tests (SVTs)
- Results in 5 min.
- Results stable for up to 60 min.
- CLIA waived
Specifications
CLIA waived | |
Human Urine | |
5 min. |