Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integra™ Miltex™ Michele Trephine
For bone cutting and biopsies
Supplier: Integra™ Miltex™ 26140
This item is not returnable.
View return policy
Description
- Surgical-grade stainless steel
- Shaft O.D. x L: 0.1875 x 6.25 in. (0.48 x 15.9cm)
- Graduated 0.25 to 1.25 in. (0.64 to 3.2cm)
Specifications
Stainless Steel | |
Michele Trephine | |
Bone Cutting and Biopsies |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title