Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integra™ Miltex™ MeisterHand Meyhoefer Chalazion Curette
Stainless steel
$384.00 - $426.00
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Price | Quantity & Availability | ||||
12-460-307
|
Integra™ Miltex™
18502 |
0.078 in. | 0.2 cm |
Each for $384.00
|
|
12-460-306
|
Integra™ Miltex™
18500 |
0.06 in. | 0.17 cm |
Each for $426.00
|
|
Description
- Length 5 in. (12.7cm)
Specifications
Stainless Steel |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title