Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Insulin, Human Recombinant, MP Biomedicals™
Insulin is a primary polypeptide hormone in humans.
Supplier: MP Biomedicals™ 0219390025
Description
Insulin is responsible for controlling the cellular uptake, utilization and storage of glucose, amino acids and fatty acids while inhibiting the breakdown of glycogen, protein and fat. It is produced by pancreatic beta cells.
- Two-chain polypeptide hormone produced by the β-cells of pancreatic islets.
- Its molecular weight is ∼5800 Da.
- The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond.
- Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.
- 1 unit corresponds to the efficiency of 0.04167mg international standard substance.
- An HPLC method exists to test the potency.
- Typical working concentration is 1 to 10μg/mL for cell culture.
Specifications
Store at –20°C. | |
Methods for immobilizing insulin on polystyrene dishes have been reported for applications involving protein-free cell cultures. | |
25 mg |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction