Learn More
Cole-Parmer Megger MIT320-EN 250V/500V/1kV Insulation Tester w/ Switch Probe
Supplier:  Cole-Parmer 2002253
Encompass_Preferred
MIT300 series insulation and continuity testers
FEATURES
  • Tough rubber armored case
  • 250V, 500V and 1000V
  • Extremely easy to use
  • Self contained protective cover
  • Live circuit warning
  • Weatherproof to IP54
  • Intelligent safety system for protection
  • USB interface
Catalog No. 50-993-490