Learn More
Cole-Parmer Megger MIT230-EN 250V/500V/1000V Insulation and Continuity Tester
Supplier:  Cole-Parmer 2002249
Encompass_Preferred
The MIT200 series insulation and continuity testers are ideal for testing transformers, motors, generators, switchgear, panel building, domestic appliances, power tools, etc., as well as fixed electrical wiring systems. Their small size and light weight make them ideal for those engineers that need to carry them for extended periods.
Catalog No. 50-993-487