Learn More
Cole-Parmer Megger MIT330-EN 250V/500V/1kV Datalogging Insulation Tester
Supplier:  Cole-Parmer 2002254
Encompass_Preferred
MIT300 series insulation and continuity testers
FEATURES
  • Tough rubber armored case
  • 250V, 500V and 1000V
  • Extremely easy to use
  • Self contained protective cover
  • Live circuit warning
  • Weatherproof to IP54
  • Intelligent safety system for protection
  • USB interface
Catalog No. 50-993-491
Explore available promotions