Learn More
Sklar Instruments Sklar Vented Instrument Protectors, Sklar 25mm, Orange, Disposable, Non-Sterile. It measures 25mm x 9.5mm x 1.5mm (1 in. x 3/8 in. x 1/16 in.). PK/100.
Supplier: Sklar Instruments 961416
SklarTip Vented Instrument Protectors, Sklar, 25mm, Orange, Disposable, Non-Sterile. Sklar Tip Protectors are used to prevent damage to instrument tips and edges. They are available in a variety of sizes and colors as well as vented or non-vented. This product is orange and vented. It measures 25mm x 9.5mm x 1.5mm (1 in. x 3/8 in. x 1/16 in.) and is sold in packs of 100.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.