Learn More
Kamiya Biomedical Insulin Calibrator, 5x0.5mL/PK RX
Supplier: Kamiya Biomedical KAI072C
K-ASSAY Insulin Calibrator. 5 x 0.5 mL (lyophilized). For 6 point calibration (purified water, not supplied, should be used as the first point). The K-ASSAY Insulin Calibrator is intended to be used for the calibration of the K-ASSAY Insulin assay (Cat. No. KAI-071). FOR IN VITRO DIAGNOSTIC USE.