Learn More
Thermo Scientific™ InSight Sensors
Thermo Scientific™ InSight sensors work with the mote transmitters, relaying data to the InSight Cloud service.
Supplier: Thermo Scientific™ INSPTCURT001
Description
Thermo Scientific™ InSight sensors relay data from the motes on your wireless account to the InSight Cloud service. In the event of internet failure, Insight Cloud will notify IT and the InSight link will log sensor data for data backfill on recovery.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
- Equipment critical parameters
- Electrical use and energy consumption
- Asset utilization
- Environmental conditions
InSight Analytics monitors your laboratory equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets’ performance against industry performance standards.
How InSight Analytics works:
- Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight web platform fpr storage.
- Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the laboratory equipment’s health to industry averages.
- This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
Specifications
Wireless Monitoring, Asset Management Solution |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Not available outside North America