Learn More
Norell Inc NORELL OPTIMIZER INSERT FOR ADAPTING 3mm TUBE TO 5mm BRUKER SPINNERS - U.S. PATENT NO. 7,728,593
Supplier: Norell Inc 5X3INSB
NORELL OPTIMIZER INSERT FOR ADAPTING 3mm TUBE TO 5mm BRUKER SPINNERS - U.S. PATENT NO. 7,728,593