Learn More
Caron Products And Services Insect Rearing Chamber, 25 cu. ft., Temp, No RH
Supplier: Caron Products And Services 7340252
Carons 25 cu. ft. (708 liters) Insect Rearing Chamber. Includes touchscreen controls, temperature range from 10C to 40C, gROD Refrigeration on Demand, and electrostatically coated refrigeration coils. Provides high volume solution for a wide variety of entomological studies. 230V, 60 Hz.