Learn More
METTLER TOLEDO InPro 6850i/12/120 Oxygen Sensor
Supplier: METTLER TOLEDO 52206119
The InPro 6850i/12/120 is an amperometric gas-phase oxygen sensor that is suited to a wide measurement range in a broad choice of applications. This sensor uses Intelligent Sensor Management (ISM) diagnostics to reduce maintenance and downtime.