Learn More
BioMed Diagnostics A self-contained system for detecting the presence of Tritrichomonas foetus. The system is simple to use and allows for collection, transport, and detection of the parasite. Available in: 10 pack
Supplier: BioMed Diagnostics 12011001
A self-contained system for detecting the presence of Tritrichomonas foetus, the parasite that causes bovine trichomoniasis. The system is simple to use and allows for collection, transport, and detection of the parasite. The proprietary medium is highly selective for T. foetus and inhibits the growth of other organisms that can interfere with a reliable diagnosis. These gold standard products provide selective media, offering several key advantages over PBS-based sample transport. | Detects: Tritrichomonas foetus | Available in: 10 pack
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.