Learn More
VEOLIA WTS Reagent, Na2S2O8, 6/pk
Supplier:  VEOLIA WTS APK 6805001
Encompass
Kit contains six pouches of dry sodium persulfate reagent. Each pouch makes 2L of 30% sodium persulfate solution.
Catalog No. NC1619637