Learn More
VEOLIA WTS Reagent, Na2S2O8, Single-pack
Supplier: VEOLIA WTS APK 6805101
Kit contains one pouch of dry sodium persulfate reagent. The pouch makes 2L of 30% sodium persulfate solution. This kit is provided with new instrument shipments.