Learn More
Innovating Science™ Water Testing Kits
Contains everything an analyst would need to perform a water analysis either in the laboratory setting or out in the field.
Supplier: Innovating Science™ IS9752
Alert:
WARNING: This product can expose you to chemicals known to the State of California to cause and reproductive harm.
Includes:
- Complete with instructions
- For 40 tests
Description
Parameters tested:
- Dissolved Oxygen
- Water Hardness
- Water's Alkalinity
- Nitrate and Nitrite
- Ammonia
Specifications
Water test Kit | |
7 to 14 | |
Determination of Water's Alkalinity | |
40 Tests |
White | |
13 to 20 | |
Instructions | |
9.75 x 6.25 x 4 in. |